ENA429358627 Enamine

1-[2-(2-fluorophenoxy)pyridin-4-yl]methanamine

  • MDL number MFCD09734469
  • Cas Number : 953886-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185744
Availability details for
Quantity
  • Availability
  • Add to Cart