ENA429358629 Enamine

4-(2-fluorophenyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09746414
  • Cas Number : 1096308-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185762
Availability details for
Quantity
  • Availability
  • Add to Cart