ENA429358632 Enamine

5-(difluoromethyl)-1-(pyridin-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD17402710
  • Cas Number : 1275366-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185785
Availability details for
Quantity
  • Availability
  • Add to Cart