ENA429358634 Enamine

3-(benzylsulfanyl)azepan-2-one

  • MDL number MFCD28125015
  • Cas Number : 88833-17-4
  • Molecular Weight : 235.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-185808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185808
Availability details for
Quantity
  • Availability
  • Add to Cart