ENA429358635 Enamine

3-(4-methoxythiophen-2-yl)-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD24051452
  • Cas Number : 1500174-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185818

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185818
Availability details for
Quantity
  • Availability
  • Add to Cart