ENA429358636 Enamine

methyl 3-(3-cyanophenyl)-1,2-oxazole-5-carboxylate

  • MDL number MFCD28118544
  • Cas Number : 1803607-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185829
Availability details for
Quantity
  • Availability
  • Add to Cart