ENA429358638 Enamine

2-cyano-N'-(3-oxobutan-2-ylidene)acetohydrazide

  • Cas Number : 119248-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185841
Availability details for
Quantity
  • Availability
  • Add to Cart