ENA429358639 Enamine

N-[5-(aminomethyl)pyridin-2-yl]acetamide

  • MDL number MFCD16737552
  • Cas Number : 1249669-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185842
Availability details for
Quantity
  • Availability
  • Add to Cart