ENA429358640 Enamine

3,4-dihydroxybenzohydrazide

  • MDL number MFCD00017064
  • Cas Number : 39635-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185843

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-185843
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart