ENA429358641 Enamine

4-methyl-3-(morpholine-4-carbonyl)aniline

  • MDL number MFCD14618163
  • Cas Number : 936210-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185845
Availability details for
Quantity
  • Availability
  • Add to Cart