ENA429358644 Enamine

3-ethyl-1-(4-methyl-3-nitrophenyl)urea

  • MDL number MFCD28118546
  • Cas Number : 1600601-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185883
Availability details for
Quantity
  • Availability
  • Add to Cart