ENA429358645 Enamine

2-{4-[(2,5-dioxoimidazolidin-4-yl)methyl]-1H-1,2,3-triazol-1-yl}acetic acid hydrochloride

  • MDL number MFCD28118548
  • Cas Number : 1786226-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185900
Availability details for
Quantity
  • Availability
  • Add to Cart