ENA429358646 Enamine

(2-methoxy-5-nitrophenyl)urea

  • MDL number MFCD28118549
  • Cas Number : 13142-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185905
Availability details for
Quantity
  • Availability
  • Add to Cart