ENA429358649 Enamine

5-chloro-1-ethyl-3-methyl-1H-pyrazole

  • MDL number MFCD04035705
  • Cas Number : 31272-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185914
Availability details for
Quantity
  • Availability
  • Add to Cart