ENA429358652 Enamine

4-(2-methoxyethoxy)cyclohexane-1-carboxylic acid

  • MDL number MFCD22418472
  • Cas Number : 1253119-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185934
Availability details for
Quantity
  • Availability
  • Add to Cart