ENA429358654 Enamine

1-hydroxy-3-methyl-6H,7H,8H,9H,10H-cyclohexa[c]chromen-6-one

  • MDL number MFCD01847420
  • Cas Number : 19815-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186360
Availability details for
Quantity
  • Availability
  • Add to Cart