ENA429358663 Enamine

3-[(dimethylamino)methylidene]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD00665175
  • Cas Number : 57315-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186369
Availability details for
Quantity
  • Availability
  • Add to Cart