ENA429358664 Enamine

4-butyl-6-chloro-7-hydroxy-2H-chromen-2-one

  • MDL number MFCD03029440
  • Cas Number : 431893-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186370

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186370
Availability details for
Quantity
  • Availability
  • Add to Cart