ENA429358673 Enamine

3-[4-(methylsulfanyl)phenyl]-1H-pyrazole

  • MDL number MFCD11558234
  • Cas Number : 474706-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186379
Availability details for
Quantity
  • Availability
  • Add to Cart