ENA429358675 Enamine

3-hydroxy-4-methyl-6H,7H,8H,9H,10H-cyclohexa[c]chromen-6-one

  • MDL number MFCD01847417
  • Cas Number : 55047-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186381
Availability details for
Quantity
  • Availability
  • Add to Cart