ENA429358676 Enamine

7,9-dihydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one

  • MDL number MFCD01325078
  • Cas Number : 83688-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186382
Availability details for
Quantity
  • Availability
  • Add to Cart