ENA429358677 Enamine

3-fluoro-7-hydroxy-4-methyl-2H-chromen-2-one

  • MDL number MFCD11108866
  • Cas Number : 97070-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186383

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186383
Availability details for
Quantity
  • Availability
  • Add to Cart