ENA429358679 Enamine

1,3-dihydroxy-6H,7H,8H,9H,10H-cyclohexa[c]chromen-6-one

  • MDL number MFCD01463067
  • Cas Number : 3722-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186385
Availability details for
Quantity
  • Availability
  • Add to Cart