ENA429358681 Enamine

5-(3,4,5-trimethoxyphenyl)-1H-pyrazole

  • MDL number MFCD11558235
  • Cas Number : 150433-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186387
Availability details for
Quantity
  • Availability
  • Add to Cart