ENA429358682 Enamine

3-(5-bromo-1H-indol-3-yl)-3-oxopropanenitrile

  • MDL number MFCD09839790
  • Cas Number : 1020722-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186388

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186388
Availability details for
Quantity
  • Availability
  • Add to Cart