ENA429358686 Enamine

1,3-dihydroxy-6H-benzo[c]chromen-6-one

  • MDL number MFCD01325155
  • Cas Number : 54245-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186392
Availability details for
Quantity
  • Availability
  • Add to Cart