ENA429358693 Enamine

2-(7-hydroxy-8-methyl-2-oxo-2H-chromen-4-yl)acetic acid

  • MDL number MFCD02676910
  • Cas Number : 68747-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186400

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186400
Availability details for
Quantity
  • Availability
  • Add to Cart