ENA429358696 Enamine

6-chloro-4-ethyl-7-hydroxy-2H-chromen-2-one

  • MDL number MFCD03029367
  • Cas Number : 143813-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186403

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186403
Availability details for
Quantity
  • Availability
  • Add to Cart