ENA429358700 Enamine

7,8-dihydroxy-3,4-dimethyl-2H-chromen-2-one

  • MDL number MFCD05997317
  • Cas Number : 1658-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186407
Availability details for
Quantity
  • Availability
  • Add to Cart