ENA429358706 Enamine

4-butyl-5-hydroxy-7-methyl-2H-chromen-2-one

  • MDL number MFCD03712848
  • Cas Number : 690680-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186413

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186413
Availability details for
Quantity
  • Availability
  • Add to Cart