ENA429358707 Enamine

7,8-dihydroxy-1H,2H,3H,4H-cyclopenta[c]chromen-4-one

  • MDL number MFCD08741891
  • Cas Number : 670275-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186414

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186414
Availability details for
Quantity
  • Availability
  • Add to Cart