ENA429358708 Enamine

6,7-dihydroxy-4-propyl-2H-chromen-2-one

  • MDL number MFCD08741836
  • Cas Number : 15941-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186415

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186415
Availability details for
Quantity
  • Availability
  • Add to Cart