ENA429358709 Enamine

4-(2-methoxyphenyl)pyrimidin-2-amine

  • MDL number MFCD07381837
  • Cas Number : 915070-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186416
Availability details for
Quantity
  • Availability
  • Add to Cart