ENA429358710 Enamine

2-[1-cyano-2-(dimethylamino)eth-1-en-1-yl]benzonitrile

  • MDL number MFCD11982954
  • Cas Number : 1144478-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186417

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186417
Availability details for
Quantity
  • Availability
  • Add to Cart