ENA429358718 Enamine

6-(3-methoxyphenyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD12143115
  • Cas Number : 76970-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186425

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186425
Availability details for
Quantity
  • Availability
  • Add to Cart