ENA429358719 Enamine

3-hydroxy-6,7-dimethoxy-2-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD18447010
  • Cas Number : 1269527-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186426

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186426
Availability details for
Quantity
  • Availability
  • Add to Cart