ENA429358723 Enamine

2-(1-oxo-1,2,5,6,7,8-hexahydrophthalazin-2-yl)acetic acid

  • MDL number MFCD16630625
  • Cas Number : 1239746-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186430
Availability details for
Quantity
  • Availability
  • Add to Cart