ENA429358731 Enamine

3-amino-6-bromo-2-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD00461730
  • Cas Number : 71822-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186438

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186438
Availability details for
Quantity
  • Availability
  • Add to Cart