ENA429358732 Enamine

2-[6-oxo-3-(thiophen-2-yl)-1,6-dihydropyridazin-1-yl]acetic acid

  • MDL number MFCD09881873
  • Cas Number : 1017326-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186439
Availability details for
Quantity
  • Availability
  • Add to Cart