ENA429358733 Enamine

3-amino-6-fluoro-2-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD02364710
  • Cas Number : 459414-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186440
Availability details for
Quantity
  • Availability
  • Add to Cart