ENA429358735 Enamine

4-(4-bromo-2-fluorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD09996850
  • Cas Number : 1010881-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186442
Availability details for
Quantity
  • Availability
  • Add to Cart