ENA429358736 Enamine

5-(1H-pyrrol-1-yl)benzene-1,3-dicarboxylic acid

  • MDL number MFCD09755869
  • Cas Number : 944894-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186443
Availability details for
Quantity
  • Availability
  • Add to Cart