ENA429358737 Enamine

4-(4-methoxyphenyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD00488391
  • Cas Number : 4875-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186444
Availability details for
Quantity
  • Availability
  • Add to Cart