ENA429358738 Enamine

5-(1H-1,2,3,4-tetrazol-1-yl)benzene-1,3-dicarboxylic acid

  • MDL number MFCD11108897
  • Cas Number : 207730-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186445
Availability details for
Quantity
  • Availability
  • Add to Cart