ENA429358740 Enamine

4-(4-chlorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD04971925
  • Cas Number : 4875-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186447
Availability details for
Quantity
  • Availability
  • Add to Cart