ENA429358746 Enamine

(4S)-2-(3-chloro-4-hydroxyphenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD15730788
  • Cas Number : 1265908-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186453
Availability details for
Quantity
  • Availability
  • Add to Cart