ENA429358751 Enamine

(4S)-2-(2,5-difluorophenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD15730785
  • Cas Number : 1272760-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186458

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186458
Availability details for
Quantity
  • Availability
  • Add to Cart