ENA429358761 Enamine

2-{3-oxo-2H,3H,5H,6H,7H,8H,9H-cyclohepta[c]pyridazin-2-yl}acetic acid

  • MDL number MFCD16170933
  • Cas Number : 1232810-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186469

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186469
Availability details for
Quantity
  • Availability
  • Add to Cart