ENA429358769 Enamine

1-(2,4-dichlorophenyl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD00172119
  • Cas Number : 137235-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186477
Availability details for
Quantity
  • Availability
  • Add to Cart