ENA429358771 Enamine

4-(2,3-dichlorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD09996849
  • Cas Number : 1010918-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186479
Availability details for
Quantity
  • Availability
  • Add to Cart